Potassium tetrafluoroborate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007226-25G 007226-500G 007226-2KG
FLUO007226-25GEA 0 GBP
FLUO007226-25G FLUO007226-500G FLUO007226-2KG
Potassium tetrafluoroborate
Potassium tetrafluoroborate
Formula: BF₄K
Boiling Pt: 960.85 °C (310 mmHg)
Melting Pt: 96.5…98 °C
Density: 3.348 g/cm³ (25 °C)
MDL Number: MFCD00011395
CAS Number: 14075-53-7
EINECS: 237-928-2
Merck Index: 12,07859

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR