Sodium hexafluoroarsenate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Sodium hexafluoroarsenate(V)

007278-10G 007278-50G
FLUO007278-10GEA 0 GBP
FLUO007278-10G FLUO007278-50G
Sodium hexafluoroarsenate
Sodium hexafluoroarsenate
Formula: AsF₆Na
MW: 211.9 g/mol
MDL Number: MFCD00014228
CAS Number: 12005-86-6
UN: 1557
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR