Potassium hexafluoroarsenate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Potassium hexafluoroarsenate(V)
002858-10G
002858-50G
002858-100G
FLUO002858-10GEA
0
GBP
FLUO002858-10G
FLUO002858-50G
FLUO002858-100G
Potassium hexafluoroarsenate
Potassium hexafluoroarsenate
Formula:
AsF₆K MW: 228.01 g/mol |
MDL Number:
MFCD00042546 CAS Number: 17029-22-0 EINECS: 241-102-7 UN: 1557 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...