Selenious acid, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
Selenous acid
M-Clarity™ quality level = MQ200
211176-10G
211176-250G
211176-50G
SIAL211176-10GEA
32.3
GBP
SIAL211176-10G
SIAL211176-250G
SIAL211176-50G
Selenious acid, Sigma-Aldrich®
Selenious acid
Formula:
H₂O₃Se Boiling Pt: 315 °C (1013 hPa) Melting Pt: 70 °C Density: 1.998 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011331 CAS Number: 7783-00-8 EINECS: 231-974-7 UN: 3283 ADR: 6.1,II Merck Index: 14,08430 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...