1,1,3,3-Tetramethylguanidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
N,N,N',N'-Tetramethylguanidine
M-Clarity™ quality level = MQ200
241768-100ML
241768-500ML
241768-5ML
SIAL241768-100MLEA
97.5
GBP
SIAL241768-100ML
SIAL241768-500ML
SIAL241768-5ML
1,1,3,3-Tetramethylguanidine, Sigma-Aldrich®
1,1,3,3-Tetramethylguanidine
Formula:
C₅H₁₃N₃ MW: 115.18 g/mol Boiling Pt: 160 Melting Pt: –30 °C Density: 0.92 g/cm³ (20 °C) Flash Pt: 54 Storage Temperature: Ambient |
MDL Number:
MFCD00008323 CAS Number: 80-70-6 EINECS: 201-302-7 UN: 2920 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...