Dibenzosuberenol 96%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Hydroxydibenzo[b,f]cycloheptene, 5H-Dibenzo[a,d][7]annulen-5-ol, 5-Dibenzosuberenol, Dibenzo[b,f]cyclohepten-1-ol, 1-Dibenzo[b,f]cycloheptenol
L02062.06
L02062.14
L02062.06EA
47.6
GBP
L02062.06
L02062.14
Dibenzosuberenol 96%
Dibenzosuberenol
Purity: 96%
CAS No.: 10354-00-4
Molecular Formula: C15H12O
Formula Weight: 208.26
Melting Point: 125-127°
Beilstein Reference: 1957458
Formula:
C₁₅H₁₂O MW: 208.26 g/mol Boiling Pt: 347 °C (1013 hPa) Melting Pt: 125…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003586 CAS Number: 10354-00-4 EINECS: 233-774-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...