2-Aminofluorene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
9H-Fluoren-2-amine, 2-Fluorenamine, Fluoren-2-ylamine
M-Clarity™ quality level = MQ100
A55500-5G
A55500-25G
SIALA55500-5GEA
58.2
GBP
SIALA55500-5G
SIALA55500-25G
2-Aminofluorene, Sigma-Aldrich®
2-Aminofluorene
Formula:
C₁₃H₁₁N MW: 181.24 g/mol Boiling Pt: 327 °C (1013 hPa) Melting Pt: 124…127 °C Density: 1.281 g/cm³ (20 °C) |
MDL Number:
MFCD00001125 CAS Number: 153-78-6 EINECS: 205-817-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...