2-Aminofluorene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 9H-Fluoren-2-amine, 2-Fluorenamine, Fluoren-2-ylamine

019454-10G 019454-50G
FLUO019454-10GEA 0 GBP
FLUO019454-10G FLUO019454-50G
2-Aminofluorene
2-Aminofluorene
Formula: C₁₃H₁₁N
MW: 181.24 g/mol
Boiling Pt: 327 °C (1013 hPa)
Melting Pt: 124…127 °C
Density: 1.281 g/cm³ (20 °C)
MDL Number: MFCD00001125
CAS Number: 153-78-6
EINECS: 205-817-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR