9,10-Phenanthrenequinone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Phenanthrene-9,10-dione
M-Clarity™ quality level = MQ100
156507-25G
156507-5G
SIAL156507-25GEA
175
GBP
SIAL156507-25G
SIAL156507-5G
9,10-Phenanthrenequinone, Sigma-Aldrich®
9,10-Phenanthrenequinone
Formula:
C₁₄H₈O₂ Boiling Pt: 360 °C (1013 hPa) Melting Pt: 203…206 °C Density: 1.4045 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001163 CAS Number: 84-11-7 EINECS: 201-515-5 UN: 3077 ADR: 9,III Merck Index: 12,07355 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...