1-Methylpiperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

022134-250G 022134-1KG
FLUO022134-250GEA 0 GBP
FLUO022134-250G FLUO022134-1KG
1-Methylpiperazine
1-Methylpiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –6 °C
Density: 0.9085 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00005966
CAS Number: 109-01-3
EINECS: 203-639-5
UN: 2734
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR