Octamethylcyclotetrasiloxane, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Volasil 244, 2,2,4,4,6,6,8,8-Octamethyl-1,3,5,7,2,4,6,8-tetraoxatetrasilocane, Silicone fluid DC 244, Dow Corning 244® fluid

M-Clarity™ quality level = MQ200

235695-100G 235695-25G
SIAL235695-100GEA 225 GBP
SIAL235695-100G SIAL235695-25G
Octamethylcyclotetrasiloxane, Sigma-Aldrich®
Octamethylcyclotetrasiloxane
Formula: [-Si(CH₃)₂O-]₄
MW: 296.62 g/mol
Boiling Pt: 175 °C (1013 hPa)
Melting Pt: 18 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00003269
CAS Number: 556-67-2
EINECS: 209-136-7
UN: 1993
ADR: 3,III
Merck Index: 12,06843

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR