Cycloheptene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
141720-25G
141720-5G
SIAL141720-25GEA
499
GBP
SIAL141720-25G
SIAL141720-5G
Cycloheptene, Sigma-Aldrich®
Cycloheptene
Formula:
C₇H₁₂ MW: 96.17 g/mol Boiling Pt: 114.6 °C (1013 hPa) Melting Pt: –56 °C Density: 0.824 g/cm³ (20 °C) |
MDL Number:
MFCD00004156 CAS Number: 628-92-2 EINECS: 211-060-4 UN: 2242 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...