cis-Cyclooctene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (Z)-Cyclooctene

M-Clarity™ quality level = MQ100

125482-25ML 125482-500ML
SIAL125482-25MLEA 46.5 GBP
SIAL125482-25ML SIAL125482-500ML
cis-Cyclooctene, Sigma-Aldrich®
cis-Cyclooctene
Formula: C₈H₁₄
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: –16 °C
Density: 0.8472 g/cm³ (20 °C)
MDL Number: MFCD00001753
CAS Number: 931-87-3
EINECS: 213-243-4
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR