3-Amino-2-cyclohexen-1-one
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
011075-5G
011075-25G
011075-100G
FLUO011075-5GEA
0
GBP
FLUO011075-5G
FLUO011075-25G
FLUO011075-100G
3-Amino-2-cyclohexen-1-one
3-Amino-2-cyclohexen-1-one
Formula:
C₆H₉NO Boiling Pt: 211 °C (1013 hPa) Melting Pt: 60 °C |
MDL Number:
MFCD00013783 CAS Number: 5220-49-5 EINECS: 226-014-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...