1,3,5,7-Cyclooctatetraene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Cyclooctatetraene, COT

M-Clarity™ quality level = MQ100

138924-1G 138924-5G
SIAL138924-1GEA 96.3 GBP
SIAL138924-1G SIAL138924-5G
1,3,5,7-Cyclooctatetraene, Sigma-Aldrich®
1,3,5,7-Cyclooctatetraene
Formula: C₈H₈
MW: 104.15 g/mol
Boiling Pt: 143
Melting Pt: –7 °C
Density: 0.925 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00004161
CAS Number: 629-20-9
EINECS: 211-080-3
UN: 2358
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR