Methylcyclopentane, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
M39407-100ML
M39407-500ML
M39407-5ML
SIALM39407-100MLEA
65.8
GBP
SIALM39407-100ML
SIALM39407-500ML
SIALM39407-5ML
Methylcyclopentane, Sigma-Aldrich®
Methylcyclopentane
Formula:
C₆H₁₂ Boiling Pt: 71.8 °C (1013 hPa) Melting Pt: –142 °C Density: 0.74 g/cm³ (20 °C) Flash Pt: –17 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001382 CAS Number: 96-37-7 EINECS: 202-503-2 UN: 2298 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...