myo-Inositol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ300

I7508-100G I7508-1KG I7508-500G I7508-50G
SIALI7508-100GEA 116 GBP
SIALI7508-100G SIALI7508-1KG SIALI7508-500G SIALI7508-50G
myo-Inositol, Sigma-Aldrich®
myo-Inositol
Formula: C₆H₁₂O₆
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 210 °C
Density: 1.752 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00077932
CAS Number: 87-89-8
EINECS: 201-781-2
Merck Index: 13,05001

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR