Cyclohexanol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Hydroxycyclohexane, Hexahydrophenol

M-Clarity™ quality level = MQ200

105899-1L 105899-2.5L 105899-25ML
SIAL105899-1LEA 44.5 GBP
SIAL105899-1L SIAL105899-2.5L SIAL105899-25ML
Cyclohexanol, Sigma-Aldrich®
Cyclohexanol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 161
Melting Pt: 25
Density: 0.947 g/cm³ (20 °C)
Flash Pt: 59
Storage Temperature: Ambient
MDL Number: MFCD00003855
CAS Number: 108-93-0
EINECS: 203-630-6
Merck Index: 13,02754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8878907005758628f5927d7b0f43a99f57de5ae8&vid=c5d64c7b5d58d205d87b42eb97190b1693d3508a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR