Cyclopropylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

125504-10G 125504-25G
SIAL125504-10GEA 82 GBP
SIAL125504-10G SIAL125504-25G
Cyclopropylamine, Sigma-Aldrich®
Cyclopropylamine
Formula: C₃H₇N
Boiling Pt: 49 °C (1013 hPa)
Melting Pt: –10 °C
Density: 0.8131 g/cm³ (20 °C)
Flash Pt: 1 °C
Storage Temperature: Ambient
MDL Number: MFCD00001301
CAS Number: 765-30-0
EINECS: 212-142-2
UN: 2733
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR