Cyclobutylamine, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Cyclobutylamine, Cyclobutanamine, Aminocyclobutane

M-Clarity™ quality level = MQ100

225185-1G 225185-25G 225185-5G
SIAL225185-1GEA 57 GBP
SIAL225185-1G SIAL225185-25G SIAL225185-5G
Cyclobutylamine, Sigma-Aldrich®
Cyclobutylamine
Formula: C₄H₉N
MW: 71.12 g/mol
Boiling Pt: 81.4
Density: 0.8328 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001328
CAS Number: 2516-34-9
EINECS: 219-736-0
UN: 2733
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR