Cyclooctyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008785-5G 008785-1G
FLUO008785-5GEA 0 GBP
FLUO008785-5G FLUO008785-1G
Cyclooctyl isothiocyanate
Cyclooctyl isothiocyanate
Formula: C₉H₁₅NS
Boiling Pt: 83…85 °C (0.8 mmHg)
Density: 1,01 g/cm³ (20 °C)
MDL Number: MFCD00040882
CAS Number: 33522-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR