Cycloheptyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
008783-5G
008783-1G
FLUO008783-5GEA
0
GBP
FLUO008783-5G
FLUO008783-1G
Cycloheptyl isothiocyanate
Cycloheptyl isothiocyanate
Formula:
C₈H₁₃NS MW: 155.26 g/mol |
MDL Number:
MFCD00041032 CAS Number: 81542-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...