Cyclobutanecarbonyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009110-10G
009110-50G
FLUO009110-10GEA
0
GBP
FLUO009110-10G
FLUO009110-50G
Cyclobutanecarbonyl chloride
Cyclobutanecarbonyl chloride
Formula:
C₅H₇ClO Boiling Pt: 60 °C (1013 hPa) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001319 CAS Number: 5006-22-4 EINECS: 225-680-8 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...