γ-Terpinene, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
gamma-Terpinene, p-Mentha-1,4-diene
M-Clarity™ quality level = MQ200
223190-5ML
223190-100ML
SIAL223190-5MLEA
33.6
GBP
SIAL223190-5ML
SIAL223190-100ML
γ-Terpinene, Sigma-Aldrich®
γ-Terpinene
Formula:
C₁₀H₁₆ MW: 136.24 g/mol Boiling Pt: 182 °C (1013 hPa) Density: 0.846 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00001537 CAS Number: 99-85-4 EINECS: 202-794-6 UN: 2319 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...