Methyl-p-benzoquinone, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Methylcyclohexa-2,5-diene-1,4-dione, Methylquinone, Methyl-1,4-benzoquinone
M-Clarity™ quality level = MQ200
211311-100G
211311-5G
SIAL211311-100GEA
280
GBP
SIAL211311-100G
SIAL211311-5G
Methyl-p-benzoquinone, Sigma-Aldrich®
Methyl-p-benzoquinone
Formula:
C₇H₆O₂ MW: 122.12 g/mol Boiling Pt: 187 °C (1013 hPa) Melting Pt: 65…67 °C Density: 1.08 g/cm³ (75.5 °C) |
MDL Number:
MFCD00001603 CAS Number: 553-97-9 EINECS: 209-056-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...