2-Methylquinoxaline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning


M-Clarity™ quality level = MQ300

W511609-1KG W511609-5KG W511609-SAMPLE
SIALW511609-1KGEA 1150 GBP
SIALW511609-1KG SIALW511609-5KG SIALW511609-SAMPLE
2-Methylquinoxaline, Sigma-Aldrich®
2-Methylquinoxaline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 245
Density: 1.11 g/cm³ (20 °C)
MDL Number: MFCD00006727
CAS Number: 7251-61-8
EINECS: 230-664-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR