2,3-Dichloroquinoxaline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
144525-10G
144525-50G
SIAL144525-10GEA
63.5
GBP
SIAL144525-10G
SIAL144525-50G
2,3-Dichloroquinoxaline, Sigma-Aldrich®
2,3-Dichloroquinoxaline
Formula:
C₈H₄Cl₂N₂ MW: 199.04 g/mol Boiling Pt: 301 °C (1013 hPa) Melting Pt: 152…153 °C |
MDL Number:
MFCD00006720 CAS Number: 2213-63-0 EINECS: 218-667-3 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...