6-Methylquinoline, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M-Clarity™ quality level = MQ100
108928-100G
108928-25G
SIAL108928-100GEA
226
GBP
SIAL108928-100G
SIAL108928-25G
6-Methylquinoline, Sigma-Aldrich®
6-Methylquinoline
Formula:
C₁₀H₉N Boiling Pt: 258.6 °C (1013 hPa) Melting Pt: –22.2 °C Density: 1.0815 g/cm³ (0 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006804 CAS Number: 91-62-3 EINECS: 202-084-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...