8-Quinolinol, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
8-Hydroxyquinoline
M-Clarity™ quality level = MQ200
H6878-100G
H6878-25G
H6878-500G
SIALH6878-100GEA
67.9
GBP
SIALH6878-100G
SIALH6878-25G
SIALH6878-500G
8-Quinolinol, Sigma-Aldrich®
8-Quinolinol
Formula:
C₉H₇NO Boiling Pt: 267 °C (1013 hPa) Melting Pt: 71.2 °C Density: 1.03 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006807 CAS Number: 148-24-3 EINECS: 205-711-1 Merck Index: 13,04869 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...