8-Aminoquinoline, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: Quinolin-8-ylamine

M-Clarity™ quality level = MQ100

260789-25G 260789-5G
SIAL260789-25GEA 198 GBP
SIAL260789-25G SIAL260789-5G
8-Aminoquinoline, Sigma-Aldrich®
8-Aminoquinoline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 290 °C (1013 hPa)
Melting Pt: 64 °C
Density: 1.337 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006809
CAS Number: 578-66-5
EINECS: 209-427-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR