4-Quinolinecarbaldehyde, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Quinoline-4-carbaldehyde, Quinoline-4-carboxaldehyde, 4-Quinolinecarboxaldehyde
M-Clarity™ quality level = MQ100
176966-10G
176966-2.5G
SIAL176966-10GEA
258
GBP
SIAL176966-10G
SIAL176966-2.5G
4-Quinolinecarbaldehyde, Sigma-Aldrich®
4-Quinolinecarbaldehyde
Formula:
C₁₀H₇NO MW: 157.17 g/mol Boiling Pt: 122…123 °C (4 mmHg) Melting Pt: 150 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006781 CAS Number: 4363-93-3 EINECS: 224-453-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...