7-Amino-4-methylcoumarin
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
7-Amino-4-methyl-2-benzopyrone, Coumarin 440, Coumarin 120, AMC
CFF561.1
CFF561.5
017134-1G
017134-25G
017134-5G
CHMPCFF561.1EA
0
GBP
CHMPCFF561.1
CHMPCFF561.5
FLUO017134-1G
FLUO017134-25G
FLUO017134-5G
7-Amino-4-methylcoumarin
7-Amino-4-methylcoumarin
Formula:
C₁₀H₉NO₂ Boiling Pt: 352 °C (1013 hPa) Melting Pt: 226 °C Density: 1,377 g/cm³ (20 °C) |
MDL Number:
MFCD00006868 CAS Number: 26093-31-2 EINECS: 247-454-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...