Hexamethylbenzene, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ100

322377-1G 322377-5G
SIAL322377-1GEA 78.6 GBP
SIAL322377-1G SIAL322377-5G
Hexamethylbenzene, Sigma-Aldrich®
Hexamethylbenzene
Formula: C₁₂H₁₈
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 82…83 °C
Density: 1.063 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008523
CAS Number: 87-85-4
EINECS: 201-777-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR