2,5-Dimethylbenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2,5-Dimethylphenyl)methanamine
018615-1G
018615-5G
FLUO018615-1GEA
0
GBP
FLUO018615-1G
FLUO018615-5G
2,5-Dimethylbenzylamine
2,5-Dimethylbenzylamine
Formula:
C₉H₁₃N MW: 135.21 g/mol |
MDL Number:
MFCD00039780 CAS Number: 93-48-1 EINECS: 202-250-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...