2,3-Dimethylbenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018613-2G 018613-5G
FLUO018613-2GEA 0 GBP
FLUO018613-2G FLUO018613-5G
2,3-Dimethylbenzylamine
2,3-Dimethylbenzylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00052680
CAS Number: 51586-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR