3,5-Bis(trifluoromethyl)benzyl alcohol
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(3,5-Bis(trifluoromethyl)phenyl)methanol
002101-5G
002101-25G
002101-100G
FLUO002101-5GEA
0
GBP
FLUO002101-5G
FLUO002101-25G
FLUO002101-100G
3,5-Bis(trifluoromethyl)benzyl alcohol
3,5-Bis(trifluoromethyl)benzyl alcohol
Formula:
C₉H₆F₆O Boiling Pt: 100 °C (14 mmHg) Melting Pt: 53…54 °C |
MDL Number:
MFCD00009907 CAS Number: 32707-89-4 EINECS: 251-168-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...