2-Chloromethyl-m-xylene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

018619-1G 018619-5G
FLUO018619-1GEA 0 GBP
FLUO018619-1G FLUO018619-5G
2-Chloromethyl-m-xylene
2-Chloromethyl-m-xylene
Formula: C₉H₁₁Cl
MDL Number: MFCD01310815
CAS Number: 5402-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR