2,4,6-Trimethylphenylboronic acid
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,4,6-Trimethylbenzeneboronic acid, (2,4,6-Trimethylphenyl)metaboric acid, Dihydroxy(2,4,6-trimethylphenyl)borane
011066-1G
011066-5G
FLUO011066-1GEA
0
GBP
FLUO011066-1G
FLUO011066-5G
2,4,6-Trimethylphenylboronic acid
2,4,6-Trimethylphenylboronic acid
Formula:
C₉H₁₃BO₂ MW: 164.01 g/mol |
MDL Number:
MFCD00236060 CAS Number: 5980-97-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...