3-Nitrobenzophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: (3-Nitrophenyl)(phenyl)methanone, m-Nitrobenzophenone

013173-25G 013173-5G
FLUO013173-25GEA 0 GBP
FLUO013173-25G FLUO013173-5G
3-Nitrobenzophenone
3-Nitrobenzophenone
Formula: C₁₃H₉NO₃
Boiling Pt: 136…138 °C (0,005 mmHg)
Melting Pt: 95 °C
Density: 1,367 g/cm³ (0 °C)
MDL Number: MFCD00007257
CAS Number: 2243-80-3
EINECS: 218-819-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR