2-(Hydroxymethyl)benzyl alcohol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: o-Phenylenedimethanol, o-Xylene-α,α'-diol, 1,2-Benzenedimethanol

M-Clarity™ quality level = MQ100

184829-10G 184829-1G
SIAL184829-10GEA 400 GBP
SIAL184829-10G SIAL184829-1G
2-(Hydroxymethyl)benzyl alcohol, Sigma-Aldrich®
2-(Hydroxymethyl)benzyl alcohol
Formula: C₈H₁₀O₂
MW: 138.17 g/mol
Boiling Pt: 291 °C (1013 hPa)
Melting Pt: 62…64 °C
MDL Number: MFCD00004626
CAS Number: 612-14-6
EINECS: 210-293-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR