2,3-Xylenol, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2,3-Dimethylphenol, 3-Hydroxy-2-xylene, 3-Hydroxy-o-xylene

M-Clarity™ quality level = MQ100

D174009-100G D174009-500G
SIALD174009-100GEA 222 GBP
SIALD174009-100G SIALD174009-500G
2,3-Xylenol, Sigma-Aldrich®
2,3-Xylenol
Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 217
Melting Pt: 75
Density: 0.965 g/cm³ (20 °C)
MDL Number: MFCD00002227
CAS Number: 526-75-0
EINECS: 208-395-3
UN: 2261
ADR: 6.1 II
Merck Index: 12,10215

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR