2,6-Xylidine, Sigma-Aldrich®
Supplier: Merck
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
M-Clarity™ quality level = MQ200
D146005-100G
D146005-500G
D146005-5G
SIALD146005-100GEA
61.9
GBP
SIALD146005-100G
SIALD146005-500G
SIALD146005-5G
2,6-Xylidine, Sigma-Aldrich®
2,6-Xylidine
Formula:
C₈H₁₁N Boiling Pt: 214 °C (1013 hPa) Melting Pt: 10…12 °C Density: 0.98 g/cm³ (20 °C) Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007747 CAS Number: 87-62-7 EINECS: 201-758-7 UN: 1711 ADR: 6.1,II Merck Index: 12,10217 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...