3-Methylbenzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: m-Tolylmethanamine, 3-(Aminomethyl)toluene

008878-1G 008878-25G 008878-5G
FLUO008878-1GEA 0 GBP
FLUO008878-1G FLUO008878-25G FLUO008878-5G
3-Methylbenzylamine
3-Methylbenzylamine
Formula: C₈H₁₁N
Boiling Pt: 202 °C (1013 hPa)
Density: 0.9809 g/cm³ (0 °C)
MDL Number: MFCD00008118
CAS Number: 100-81-2
EINECS: 202-890-8
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR