2,5-Xylyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2,5-Dimethylphenylisothiocyanate, 2,5-Dimethylphenyl isothiocyanate
002552-5G
002552-25G
FLUO002552-5GEA
0
GBP
FLUO002552-5G
FLUO002552-25G
2,5-Xylyl isothiocyanate
2,5-Xylyl isothiocyanate
Formula:
C₉H₉NS MW: 163.24 g/mol |
MDL Number:
MFCD00046798 CAS Number: 19241-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...