3,5-Xylyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,5-Dimethylphenyl isothiocyanate, 3,5-Dimethylphenylisothiocyanate

008924-5G 008924-25G
FLUO008924-5GEA 0 GBP
FLUO008924-5G FLUO008924-25G
3,5-Xylyl isothiocyanate
3,5-Xylyl isothiocyanate
Formula: C₉H₉NS
MW: 163.24 g/mol
Boiling Pt: 261 °C (1013 hPa)
MDL Number: MFCD00041082
CAS Number: 40046-30-8
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR