2,4-Xylyl isothiocyanate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002551-5G
002551-25G
FLUO002551-5GEA
0
GBP
FLUO002551-5G
FLUO002551-25G
2,4-Xylyl isothiocyanate
2,4-Xylyl isothiocyanate
Formula:
C₉H₉NS Boiling Pt: 262…263 °C (1013 hPa) Melting Pt: 31.5 °C |
MDL Number:
MFCD00022054 CAS Number: 39842-01-8 UN: 2923 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...