2,6-Xylyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,6-Dimethylphenylisothiocyanate, 2,6-Dimethylphenyl isothiocyanate

002553-5G 002553-25G 002553-100G
FLUO002553-5GEA 0 GBP
FLUO002553-5G FLUO002553-25G FLUO002553-100G
2,6-Xylyl isothiocyanate
2,6-Xylyl isothiocyanate
Formula: C₉H₉NS
Boiling Pt: 67 °C (0.4 mmHg)
Density: 1.085 g/cm³ (20 °C)
MDL Number: MFCD00038715
CAS Number: 19241-16-8
EINECS: 242-906-0
UN: 3334
ADR: 9,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR