3,4-Xylyl isothiocyanate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Dimethylphenyl isothiocyanate

002554-5G 002554-25G 002554-1G
FLUO002554-5GEA 0 GBP
FLUO002554-5G FLUO002554-25G FLUO002554-1G
3,4-Xylyl isothiocyanate
3,4-Xylyl isothiocyanate
Formula: C₉H₉NS
MW: 163.24 g/mol
MDL Number: MFCD00031536
CAS Number: 19241-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR