2,3-Xylyl isothiocyanate

Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,3-Dimethylphenyl isothiocyanate

D5660.10 D5660.50
XXXXD5660.10EA 0 GBP
XXXXD5660.10 XXXXD5660.50
2,3-Xylyl isothiocyanate
2,3-Xylyl isothiocyanate
Formula: C₉H₉NS
MW: 163.24 g/mol
MDL Number: MFCD00041064
CAS Number: 1539-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR