5-Iodo-m-xylene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Iodo-3,5-dimethylbenzene, 3,5-Dimethyl-1-iodobenzene, 1,3-Dimethyl-5-iodobenzene, 3,5-Dimethyliodobenzene
002953-25G
002953-100G
FLUO002953-25GEA
0
GBP
FLUO002953-25G
FLUO002953-100G
5-Iodo-m-xylene
5-Iodo-m-xylene
Formula:
C₈H₉I MW: 232.06 g/mol Boiling Pt: 234…235 °C (1013 hPa) Density: 1,61 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00060659 CAS Number: 22445-41-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...